Learn More
Andwin Scientific Adhesive PCR Film Seals, Nonsterile, 100/cs
SDP
Catalog No. NC1397036
Encompass_Preferred
These heat resistant 50 um thick films are designed for thermal cycling applications. Polypropylene films are not pierceable. For PCR applications, where piercing with pipette tips or robotic probes is required for product recovery, use aluminium foil films. For Real-Time PCR applications, where maximum optical clarity is required, use optically-clear polyester films.
Catalog No. NC1397036 Supplier Andwin Scientific Supplier No. 60941070
These heat resistant 50 um thick films are designed for thermal cycling applications. Polypropylene films are not pierceable. For PCR applications, where piercing with pipette tips or robotic probes is required for product recovery, use aluminium foil films. For Real-Time PCR applications, where maximum optical clarity is required, use optically-clear polyester films.