Learn More
Hach Company Hardness Quality Control Standard, High Range, 500 mL
Supplier:  Hach Company 2833349
Encompass
Ready to use - no dilutions required. Mixed standard includes: Total Hardness: 1000 mg/L as CaCO₃, Calcium Hardness: 500 mg/L as CaCO₃. 500 mL Bottle
Catalog No. 50-199-6738