Learn More
Hach Company Hypochlorite Test Kit, Model CN-HRDT
Supplier:  Hach Company 2687100
Encompass
Utilizing the Hach Digital Titrator, this kit is used to determine hypochlorite, high range (Bleach) solution concentrations. Kit includes Digital Titrator, reagents and labware, manual and carrying case. Range: 5 - 15% as Cl2: 50 - 150 g/L as Cl2. Contains 100 tests.
Catalog No. 50-199-6545