Learn More
Hach Company HR Manganese Chemkey
Supplier:  Hach Company 3007000
Encompass
HR Manganese Chemkey Reagents (box of 25)For determination of Manganese using Hach Method 10319Ensure SL1000/SL250 has been updated with Software Version V3 0 12 1489 to use this Chemkey
Catalog No. 50-002-07005