Learn More
Hach Company Nitrite TNTplus Vial Test, HR (0.6-6.0 mg/L NO₂-N), 25 Tests
Catalog No. 501998133 Shop All Hach Company Products
Encompass
Nitrite TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Nitrite Nitrogen by the Diazotization method. EPA compliant.
Catalog No. 50-199-8133 Supplier Hach Company Supplier No. TNT840
Nitrite TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Nitrite Nitrogen by the Diazotization method. EPA compliant.