Learn More
TA Instruments HR x0, Stainless Steel, Sandblasted, 25 mm Plate, NST, Upper Plate for Use with Lower Peltier Plate, Smart Swap 2 Geometry, Surface Finish 80–84 RMS or Ra 1.8–2.0 µm, Compatible with HR x0 Instruments Only
Catalog No. 5000580821 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR x0, Stainless Steel, Sandblasted, 25 mm Plate, NST is an upper plate for use with lower Peltier plate. It features Smart Swap 2 geometry for use with HR x0 instruments only. Surface finish is 80–84 RMS in microinches or Ra value of 1.8 to 2.0 microns.
Catalog No. 50-005-80821 Supplier TA Instruments Supplier No. 519250.946
Waters TA Instruments HR x0, Stainless Steel, Sandblasted, 25 mm Plate, NST is an upper plate for use with lower Peltier plate. It features Smart Swap 2 geometry for use with HR x0 instruments only. Surface finish is 80–84 RMS in microinches or Ra value of 1.8 to 2.0 microns.