Learn More
Hampton Research Detergent Screen, 0.25 mL, Deep Well block format
SDP
Catalog No. NC1464149
Encompass
Manipulate sample-sample and sample-solvent interactions, including non-specific aggregation due to hydrophobic interactions, to alter sample solubility, or obtain or improve crystalline samples.
Catalog No. NC1464149 Supplier Hampton Research Supplier No. HR2407
Manipulate sample-sample and sample-solvent interactions, including non-specific aggregation due to hydrophobic interactions, to alter sample solubility, or obtain or improve crystalline samples.