Learn More
Medchemexpress LLC Hr97 Tfa | 1518.84 | 1 MG
SDP
Catalog No. 5000422954
Encompass_Preferred
HR97 TFA is a cell-penetrating peptide that can be combined with engineered melanin to prepare eye drops called HR97-SunitiGel. The peptide-drug conjugate can provide sustained ocular drug delivery.
  • Cell-penetrating peptide
  • Combines with engineered melanin to prepare eye drops
  • Supports sustained ocular drug delivery
  • Sequence: Phe-Ser-Gly-Lys-Arg-Arg-Lys-Arg-Lys-Pro-Arg-Cys
  • Shortened sequence: FSGKRRKRKPRC
  • In vitro solubility: 50 mg/mL in DMSO (need ultrasonic)
Catalog No. 50-004-22954 Supplier Medchemexpress LLC Supplier No. HYP108951MG
HR97 TFA is a cell-penetrating peptide that can be combined with engineered melanin to prepare eye drops called HR97-SunitiGel. The peptide-drug conjugate can provide sustained ocular drug delivery.
  • Cell-penetrating peptide
  • Combines with engineered melanin to prepare eye drops
  • Supports sustained ocular drug delivery
  • Sequence: Phe-Ser-Gly-Lys-Arg-Arg-Lys-Arg-Lys-Pro-Arg-Cys
  • Shortened sequence: FSGKRRKRKPRC
  • In vitro solubility: 50 mg/mL in DMSO (need ultrasonic)