Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HR97 TFA | 1518.84 | 5 MG
SDP

Catalog No. 5000422952
Encompass_Preferred

HR97 TFA is a cell-penetrating peptide that can be combined with engineered melanin to prepare eye drops HR97-SunitiGel. The peptide-drug conjugate in HR97-SunitiGel can provide sustained ocular drug delivery.

  • Cell-penetrating peptide
  • Molecular weight of 1518.84 (free acid)
  • Sequence: Phe-Ser-Gly-Lys-Arg-Arg-Lys-Arg-Lys-Pro-Arg-Cys
  • Solubility in DMSO: 50 mg/mL (requires ultrasonic)
  • Can be used to prepare HR97-SunitiGel eye drops

Catalog No. 50-004-22952 Supplier Medchemexpress LLC Supplier No. HYP108955MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HR97 TFA is a cell-penetrating peptide that can be combined with engineered melanin to prepare eye drops HR97-SunitiGel. The peptide-drug conjugate in HR97-SunitiGel can provide sustained ocular drug delivery.

  • Cell-penetrating peptide
  • Molecular weight of 1518.84 (free acid)
  • Sequence: Phe-Ser-Gly-Lys-Arg-Arg-Lys-Arg-Lys-Pro-Arg-Cys
  • Solubility in DMSO: 50 mg/mL (requires ultrasonic)
  • Can be used to prepare HR97-SunitiGel eye drops

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.