Learn More
Sino Biological Recombinant Human GNAZ Protein (GST Tag)
Catalog No. 502574797
Encompass_Preferred
Recombinant human GNAZ (2-end) protein was expressed in E.coli cells using an N-terminal GST tag.
Catalog No. 50-257-4797 Supplier Sino Biological Supplier No. G13130G100
Recombinant human GNAZ (2-end) protein was expressed in E.coli cells using an N-terminal GST tag.