Learn More
Aviva Systems Biology Corp HRG Antibody - middle region (ARP41397_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41397_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HRG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HRG. Alternate names: DKFZp779H1622, HPRG, HRGPApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-014-57831