Learn More
Aviva Systems Biology Corp HRH2 Antibody - C-terminal region (ARP65575_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65575_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HRH2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human HRH2. Alternate names: H2RApplicationWB. Species reactivity: Dog, Horse, Human, Rat
Catalog No. 50-288-523