Learn More
Eurofins Discoverx HRI, active, 250 µg. (16-013M)
Supplier:  Eurofins Discoverx 16013M
Encompass
N-terminal GST-tagged recombinant, human HRI amino acids 140-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9519