Learn More
Carnabio Usa Inc HRI(EIF2AK1)/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0515410
Encompass
HRI(EIF2AK1), Wild type, Amino Acid 1-630(end), BAC11461.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1121