Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter HRLD-400 SENSOR

Catalog No. NC2459091
Encompass

  • Compatible with LPC Instruments platform.
  • Analyzes particle size range from 2.0 to 400 µm.
  • Supports flow rates between 20 and 100 mL/min.
  • Utilizes light obscuration measurement method.
  • Employs a Class III B laser diode as the light source.
  • Features a laser wavelength of 780 nm.
  • Operates with a laser power of 5 mW.
  • Handles maximum particle number concentration of 10,000/mL.
  • Supports maximum sample temperature up to 65 °C.
  • Compatible with multiple solvents, including hydraulic oil and purified water.
  • Designed for indoor use only.
  • Weighs 0.5 kg.
  • Measures 203.2 mm in width, 38.1 mm in depth, and 38.1 mm in height.

Catalog No. NC2459091 Supplier Beckman Coulter Supplier No. 2089250400

To request a quote for this item, please select Request a Quote to reach our dedicated support team.

missing translation for 'validMainframeMsg'

  • Compatible with LPC Instruments platform.
  • Analyzes particle size range from 2.0 to 400 µm.
  • Supports flow rates between 20 and 100 mL/min.
  • Utilizes light obscuration measurement method.
  • Employs a Class III B laser diode as the light source.
  • Features a laser wavelength of 780 nm.
  • Operates with a laser power of 5 mW.
  • Handles maximum particle number concentration of 10,000/mL.
  • Supports maximum sample temperature up to 65 °C.
  • Compatible with multiple solvents, including hydraulic oil and purified water.
  • Designed for indoor use only.
  • Weighs 0.5 kg.
  • Measures 203.2 mm in width, 38.1 mm in depth, and 38.1 mm in height.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.