Learn More
Biolegend HRP anti-β-Amyloid, 1-42
Supplier:  Biolegend 805507
Encompass_Preferred
HRP anti-β-Amyloid, 1-42 [12F4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB; Size: 200 uL
Catalog No. 50-207-3945