Learn More
Immunology Consultants HRP Conjugated Chicken anti-Cotton Rat IgG h+l Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants CCOT25P
Encompass_Preferred
Chickens were immunized with highly purified Cotton rat IgG and eggs were collected. Antibody was sold phase adsorbed to remove cross reactive antibody. Specific antibody was immunoaffinity purified off a Cotton Rat IgG containing immunosorbent and conjugated with horseradish peroxidase (HRP). Antibody concentration was determined prior to conjugation using an absorbance at 280 nm: 1.32 equals 1.0 mg of IgY. This antibody reacts with Cotton Rat IgG as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA, and IHC applications. This antibody may cross-react with IgG from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-224-8870