Learn More
Biolegend HRP anti-His Tag
Supplier:  Biolegend 652503
Encompass_Preferred
HRP anti-His Tag [J099B12]; Isotype: Mouse IgG1, κ; Reactivity: All species (6x His tag epitope); Apps: ELISA, WB; Size: 25 ug
Catalog No. NC1737899