Learn More
Biolegend HRP anti-human β2-microglobulin
Supplier:  Biolegend 280303
Encompass_Preferred
HRP anti-human β2-microglobulin [2M2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: ELISA Detection, FC; Size: 50 ug
Catalog No. 50-113-0012