Learn More
LifeSensors HRP-labeled Anti-SUMO/SUMOstar Antibody 50µg
SDP
Supplier:  LifeSensors AB70020051
Encompass_Preferred
Our Anti-SUMO/SUMOstar antibody is an affinity-purified, polyclonal chicken IgY and labeled with horseradish peroxidase. It can be used for simplified detection of yeast SUMO- and SUMOstar-tagged polypetides by Western blot and ELISA. This antibody is HRP-labeled
Catalog No. 50-103-7504