Learn More
Abcam HRP Conjugation Kit - Lightning-Link, 1 x 5MG
Supplier:  Abcam AB1028901X5MG
Encompass_Preferred
Abcam's HRP Conjugation Kit (ab102890): Fast, Efficient, and Reliable - Rapid Conjugation: Achieve HRP conjugation in under 4 hours with just 30 seconds of hands-on time using the HRP Conjugation Kit ab102890. - High Efficiency: Ensures 100% antibody recovery, preserving the integrity and value of your antibodies. - Effortless Protocol: Add modifier to your antibody, incubate for 3 hours, then add quencher for 30 minutes, with no need for further purification. - Scalable: Compatible with most standard antibody formulations, effective for 10µg to 100mg of antibody. - Versatile Conjugation: Utilize Lightning-Link technology to conjugate with over 45 labels, including Alexa Fluor, DyLight, tandem dyes, enzymes, oligonucleotides, and gold nanoparticles. - Confidence: cited in over 60 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2077083