Learn More
Electron Microscopy Sciences IHC Retriever HRP Conjugate Diluent 125 ML
SDP
Catalog No. 501926540
Encompass_Preferred
Specifically designed for preparing solution of your HRP-conjugate used as the detection reagent. It is the Antibody-dilutent buffer with additional component for stabilizing your HRP-conjugate. Allows you to further standardize the assay preparing ready-to-use conjugate solutions in advance and store them in refrigerator without loss of activity.
Catalog No. 50-192-6540 Supplier Electron Microscopy Sciences Supplier No. 62717
Specifically designed for preparing solution of your HRP-conjugate used as the detection reagent. It is the Antibody-dilutent buffer with additional component for stabilizing your HRP-conjugate. Allows you to further standardize the assay preparing ready-to-use conjugate solutions in advance and store them in refrigerator without loss of activity.