Learn More
Abcam HRP Conjugate Stabilizer - Life x tend, 50ML
Catalog No. 502656162
Encompass_Preferred
HRP Conjugate Stabilizer - LifeXtend (ab270548) is a proprietary multi-component reagent system that protects antibody–HRP conjugates from instability, microbrial attack, and denaturation, thus ensuring the best possible performance in experiments performed at room temperature.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6162 Supplier Abcam Supplier No. AB27054850ML
HRP Conjugate Stabilizer - LifeXtend (ab270548) is a proprietary multi-component reagent system that protects antibody–HRP conjugates from instability, microbrial attack, and denaturation, thus ensuring the best possible performance in experiments performed at room temperature.
The product is subject to the following: Abcam Restricted Use Statement