Learn More
Bio Rad Laboratories HRP CONJUGATE SUBSTRATE KIT
Catalog No. 509010989
Encompass
Kit, Substrate; Bio-Rad; HRP Conjugate; Contains premixed reagents; 4-chloro-1-naphthol, hydrogen peroxide solutions, color development buffer; makes 1 L color development solution
Catalog No. 50-901-0989 Supplier Bio Rad Laboratories Supplier No. 1706431
Kit, Substrate; Bio-Rad; HRP Conjugate; Contains premixed reagents; 4-chloro-1-naphthol, hydrogen peroxide solutions, color development buffer; makes 1 L color development solution