Learn More
ABclonal Technology HRP-conjugated B-Actin Mouse
SDP
Supplier:  ABclonal Technology AC043100UL
Encompass_Preferred
This gene encodes one of six different actin proteins Actins are highly conserved proteins that are involved in cell motility structure integrity and intercellular signaling The encoded protein is a major constituent of the contractile apparatus and one of the two nonmuscle cytoskeletal actins that are ubiquitously expressed Mutations in this gene cause Baraitser-Winter syndrome 1 which is characterized by intellectual disability with a distinctive facial appearance in human patients Numerous pseudogenes of this gene have been identified throughout the human genome
Catalog No. 50-000-13824