Learn More
Proteintech Group Inc HRP-conjugated Beta Actin Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc HRP66009100UL
Encompass_Preferred
The Beta Actin antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Beta Actin. This antibody recognizes human, mouse, rat, hamster, zebrafish, monkey antigen. The Beta Actin antibody has been validated for the following applications: WB analysis. This antibody is hrp.
Catalog No. 50-225-8356