Learn More
Medchemexpress LLC HRP-conjugated Donkey Anti-Rabbit IgG H&L | 100 UL
SDP
Catalog No. 5000449715
Encompass_Preferred
HRP-conjugated Donkey Anti-Rabbit IgG H&L is a HRP-conjugated and Donkey origined Polyclonal antibody, targeting to Rabbit IgG. Donkey Anti-Rabbit IgG H&L(HRP) can binds to the light and heavy chains of Rabbit IgG antibodies.
  • WB (Western Blot): 1:5000 - 1:20000
  • IHC-P (Immunohistochemistry-Paraffin): 1:500 - 1:1000
  • ICC/IF (Immunocytochemistry/Immunofluorescence): 1:500 - 1:1000
  • ELISA (Enzyme Linked Immunosorbent Assay): 1:5000 - 1:20000
Catalog No. 50-004-49715 Supplier Medchemexpress LLC Supplier No. HYP84939100UL
HRP-conjugated Donkey Anti-Rabbit IgG H&L is a HRP-conjugated and Donkey origined Polyclonal antibody, targeting to Rabbit IgG. Donkey Anti-Rabbit IgG H&L(HRP) can binds to the light and heavy chains of Rabbit IgG antibodies.
  • WB (Western Blot): 1:5000 - 1:20000
  • IHC-P (Immunohistochemistry-Paraffin): 1:500 - 1:1000
  • ICC/IF (Immunocytochemistry/Immunofluorescence): 1:500 - 1:1000
  • ELISA (Enzyme Linked Immunosorbent Assay): 1:5000 - 1:20000