Learn More
Proteintech Group Inc HRP-conjugated GST Tag Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc HRP66001100UL
Encompass_Preferred
The GST Tag antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of GST Tag. This antibody recognizes recombinant protein, schistosoma japonicum antigen. The GST Tag antibody has been validated for the following applications: WB analysis. This antibody is hrp.
Catalog No. 50-225-8354