Learn More
ACROBiosystems ACROBiosystems HRP-Conjugated Monoclonal Mouse Anti-DXD & Exatecan Antibody, IgG1, 200 μg (DXD-PLM684-200ug)
Catalog No. NC2745977
Encompass_Preferred
HRP-conjugated mouse monoclonal antibody specific for DXD and exatecan, formatted for immunoassay applications including ELISA and PK/PD studies. Supplied lyophilized and purified to support reproducible detection in research assays.
  • HRP conjugated for enzyme-based detection.
  • Mouse IgG1 isotype.
  • Supplied lyophilized from 0.22 μm filtered PBS with trehalose protectant.
  • Protein A/G purified.
  • Recommended ELISA working range 2-5,000 ng/mL.
  • Store lyophilized at -20°C or lower; after reconstitution store at -70°C and avoid repeated freeze-thaw cycles.
Catalog No. NC2745977 Supplier ACROBiosystems Supplier No. DXDPLM684200UG
HRP-conjugated mouse monoclonal antibody specific for DXD and exatecan, formatted for immunoassay applications including ELISA and PK/PD studies. Supplied lyophilized and purified to support reproducible detection in research assays.
  • HRP conjugated for enzyme-based detection.
  • Mouse IgG1 isotype.
  • Supplied lyophilized from 0.22 μm filtered PBS with trehalose protectant.
  • Protein A/G purified.
  • Recommended ELISA working range 2-5,000 ng/mL.
  • Store lyophilized at -20°C or lower; after reconstitution store at -70°C and avoid repeated freeze-thaw cycles.