Learn More
Medchemexpress LLC Hrp-conjugated rabbit anti-rat IgG H&L | 50 UL
SDP
Catalog No. 5000443692
Encompass_Preferred
HRP-conjugated Rabbit Anti-Rat IgG H&L is a HRP-conjugated rabbit-originated antibody targeting the rat IgG antibody. It is isolated as an intact whole IgG molecule from antisera by immunoaffinity chromatography, featuring an Fc portion and two antigen binding Fab portions. It is suitable for various applications including Western Blot, Immunohistochemistry-Paraffin, and ELISA.
  • Targeted to rat IgG antibody
  • Isolated as intact whole IgG molecules
  • Suitable for Western Blot, IHC-P, and ELISA applications
  • Supplied in phosphate buffered saline with BSA and glycerol
Catalog No. 50-004-43692 Supplier Medchemexpress LLC Supplier No. HYP8373850UL
HRP-conjugated Rabbit Anti-Rat IgG H&L is a HRP-conjugated rabbit-originated antibody targeting the rat IgG antibody. It is isolated as an intact whole IgG molecule from antisera by immunoaffinity chromatography, featuring an Fc portion and two antigen binding Fab portions. It is suitable for various applications including Western Blot, Immunohistochemistry-Paraffin, and ELISA.
  • Targeted to rat IgG antibody
  • Isolated as intact whole IgG molecules
  • Suitable for Western Blot, IHC-P, and ELISA applications
  • Supplied in phosphate buffered saline with BSA and glycerol