Learn More
Vector Laboratories Streptavidin, Horseradish Peroxidase Conjugated, Concentrate, For ELISAs And Blots 1 mg
SDP
Catalog No. NC0665354
Encompass
Vector Laboratories streptavidin and avidin reagents are highly purified and possess very low non-specific binding properties. They have a high affinity for biotin and can be used to detect biotinylated secondary antibodies and other macromolecules. Horseradish Peroxidase Streptavidin (HRP Streptavidin) is produced by a coupling procedure that preserves the high specific activity of the peroxidase. This concentrated HRP Streptavidin is specifically formulated for use in ELISAs and blots.
Catalog No. NC0665354 Supplier Vector Laboratories Supplier No. SA50141
Vector Laboratories streptavidin and avidin reagents are highly purified and possess very low non-specific binding properties. They have a high affinity for biotin and can be used to detect biotinylated secondary antibodies and other macromolecules. Horseradish Peroxidase Streptavidin (HRP Streptavidin) is produced by a coupling procedure that preserves the high specific activity of the peroxidase. This concentrated HRP Streptavidin is specifically formulated for use in ELISAs and blots.