Learn More
Abcam Rabbit specific HRP/DAB Detection IHC Detection Kit - Micro-polymer, 15ML
Catalog No. NC1872120
Encompass_Preferred
This IHC kit is used to detect rabbit primary antibodies in immunohistochemistry (IHC).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1872120 Supplier Abcam Supplier No. AB23646915ML
This IHC kit is used to detect rabbit primary antibodies in immunohistochemistry (IHC).
The product is subject to the following: Abcam Restricted Use Statement