Learn More
Seracare Life Sciences Inc HRP ELISA KIT ANTI-HUMAN
Supplier:  Seracare Life Sciences Inc 51100009
Encompass_Preferred
Highly sensitive reagents optimized for antigen detection in picogram quantities. Pretested wash and blocking solutions ensure low background. Can be used for both direct and indirect sandwich assays. Protein Detector ELISA Kits offer a convenient starting point for the development of microwell ELISA protocols for antigen and antibody detection. These kits provide pretested components and suggested protocols to facilitate the development of ELISA procedures. KPL's protein Detector ELISA Kit, HRP Systems are provided with ABTS Peroxidase Substrate which offers performance, reliability, and low background.
Catalog No. 50-675-11