Learn More
Seracare Life Sciences Inc HRP ELISPOT KIT
Supplier:  Seracare Life Sciences Inc 55300005
Encompass
Kit offers pretested reagents and a convenient, optimized protocol for ELISpot protein detection via HRP-labeled streptavidin. With all the necessary reagents except the paired antibody set, this kit allows for minimal time and effort in developing a sensitive and reproducible ELISpot assay. Plates and reagents are provided for the detection of 5 plates.
Catalog No. 50-673-83