Learn More
Immunology Consultants HRP Conjugated Goat anti-Mouse C3 Antibody, 0.1 mg
SDP
Supplier:  Immunology Consultants GC390PZ
Encompass_Preferred
Goats were immunized with highly purified mouse C3 and the resulting antiserum was collected. Antibody was immunoaffinity purified off a mouse C3 containing immunosorbent and conjugated with horseradish peroxidase (HRP). Antibody concentration was determined prior to conjugation using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody will react with mouse C3 as determined by ELISA and IEP techniques. It is suitable for blotting, ELISA and IHC applications. This antibody may cross-react with C3 from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-224-8792