Learn More
Active Motif HRP Goat anti-Rabbit IgG
SDP
Supplier:  Active Motif 15015
Encompass
Active Motif's high quality anti-rabbit IgG secondary antibody was raised in goat and has been affinity purified and conjugated to horseradish peroxidase (HRP). By improving this procedure we are able to offer antibodies with high specificity, which will reduce the background and increase the sensitivity of detection in your assay.
Catalog No. NC1223863