Learn More
LICOR WesternSure Goat anti-Mouse HRP Secondary Antibody (1 mL)
Supplier:  LICOR 92680010
Encompass_Preferred
Immunogen: Mouse IgG paraproteins | Purity and Specificity: The antibody was purified by affinity chromatography on pooled mouse IgG covalently linked to agarose. Based on ELISA and/or flow cytometry, the antibody reacts with the heavy and light chains of mouse IgG1, IgG2a, IgG2b, and IgG3, along with the light chains of mouse IgM and IgA. The antibody has minimal cross-reactivity with human immunoglobulins. | Application: WesternSure Goat anti-Mouse HRP Secondary Antibody is recommended for Chemiluminescent Western blots | Formulation and Recommended Dilutions: The antibody is supplied as purified immunoglobulin, supplied in 50% glycerol and 50% PBS, pH 7.4. No preservative has been added. Store at 4 °C.
Catalog No. 50-489-553