Learn More
LICOR WesternSure Goat anti-Rabbit HRP Secondary Antibody (1 mL)
Supplier:  LICOR 92680011
Encompass_Preferred
Immunogen: Rabbit IgG | Application: WesternSure Goat anti-Rabbit HRP Secondary Antibody is recommended for Chemiluminescent Western blots | Formulation and Recommended Dilutions: The antibody is supplied as purified immunoglobulin, supplied in 50% glycerol and 50% PBS, pH 7.4. No preservative has been added. Store at 4 °C.
Catalog No. 50-489-554