Learn More
Seracare Life Sciences Inc HRP-LABELED STREPTAVIDIN
Supplier:  Seracare Life Sciences Inc 59500004
Encompass_Preferred
Streptavidin labeled with horseradish peroxidase. Streptavidin is a protein isolated from the bacterium Streptomyces avidinii and has no carbohydrate moieties. It has a molecular weight of approximately 60,000 Daltons with an isoelectric point of 5. Product is in liquid form. Peroxidase labeled streptavidin is ideally suited ELISA, blotting, immunohistochemistry, and in situ hybridization applications.
Catalog No. 50-673-92