Learn More
Vector Laboratories ImmPRESS Excel Amplified HRP Polymer Staining Kit (Anti-Mouse IgG), Made In Horse 15 ml
SDP
Supplier:  Vector Laboratories MP760215
Encompass
The ImmPRESS Excel Amplified HRP (Peroxidase) Polymer Staining Kit (Anti-Mouse IgG) includes an enzymatic amplification system that produces crisp, highly sensitive, specific staining with low background. This system employs a proprietary ready-to-use (RTU) Amplifier Antibody, followed by an RTU ImmPRESS Excel HRP Polymer Reagent. These reagents are affinity purified and extensively cross-adsorbed, and concentrate a higher density of reporter enzyme on the target with minimal interference. The included ImmPACT DAB EqV Substrate produces a crisp, dark brown reaction product with excellent sensitivity and low background. The kit contains: BLOXALL Endogenous Enzyme Blocking Solution (RTU), 2.5% Normal Horse Serum (RTU), Amplifier Antibody (RTU), ImmPRESS Excel Polymer Reagent (RTU), ImmPACT DAB EqV. Stains 75-150 sections
Catalog No. NC0695806