Learn More
Boca Scientific Inc HRP-Protector™, 125ml
Catalog No. NC1894499
Encompass
HRP-Protector™ is a stabilization reagent for horseradish peroxidase (HRP) coupled to antibodies or Neutravidin/Streptavidin. It provides long-term stability over several years even when stored in low concentrated, ready-to-use dilutions. HRP-Protector™ can be used directly as an assay buffer for incubating the HRP-conjugates.
Catalog No. NC1894499 Supplier Boca Scientific Inc Supplier No. 222 125
HRP-Protector™ is a stabilization reagent for horseradish peroxidase (HRP) coupled to antibodies or Neutravidin/Streptavidin. It provides long-term stability over several years even when stored in low concentrated, ready-to-use dilutions. HRP-Protector™ can be used directly as an assay buffer for incubating the HRP-conjugates.