Learn More
Genscript Corporation Antibody Protein Protein Analysis Protein Analysis 1 MG 1/EA
Supplier:  Genscript Corporation M00098
Encompass
Protein L is a cell surface protein expressed by Peptostreptoccocus magnus that binds to the variable light chains (kappa chains) of immunoglobulins without interfering with antigen binding. In contrast to IgG-binding proteins such as protein A and protein G protein L can be used for the detection and purification of mammalian kappa light chain antibodies of all classes. HRP-Protein L is produced by covalently linking HRP to protein L. The complex can be used in immuno-detection such as immunoblotting (western or dot blot) ELISA and immunohistochemistry.
Catalog No. 50-252-9964