Learn More
Apexbio Technology LLC HRP Rabbit Anti-Goat IgG (H+L) Antibody 1ml
SDP
Catalog No. 5000097035
Encompass_Preferred
HRP Rabbit Anti-Goat IgG HplusL Antibody is a polyclonal secondary antibody targeting goat immunoglobulins (IgG heavy and light chains) It is designed to bind specifically to goat-derived primary antibodies enabling enzymatic signal amplification HRP Rabbit Anti-Goat IgG HplusL Antibody exerts its biological activity through horseradish peroxidase (HRP) conjugation which catalyzes chromogenic substrate conversion for colorimetric or chemiluminescent detection Based on these properties HRP Rabbit Anti-Goat IgG HplusL Antibody is applicable in Western blotting ELISA immunohistochemistry and immunocytochemistry
Catalog No. 50-000-97035 Supplier Apexbio Technology LLC Supplier No. K12241
HRP Rabbit Anti-Goat IgG HplusL Antibody is a polyclonal secondary antibody targeting goat immunoglobulins (IgG heavy and light chains) It is designed to bind specifically to goat-derived primary antibodies enabling enzymatic signal amplification HRP Rabbit Anti-Goat IgG HplusL Antibody exerts its biological activity through horseradish peroxidase (HRP) conjugation which catalyzes chromogenic substrate conversion for colorimetric or chemiluminescent detection Based on these properties HRP Rabbit Anti-Goat IgG HplusL Antibody is applicable in Western blotting ELISA immunohistochemistry and immunocytochemistry