Learn More
Seracare Life Sciences Inc HRP-RABBIT ANTI-YERSINIA
Supplier:  Seracare Life Sciences Inc 53200030
Encompass_Preferred
Protein G purified BacTrace antibody to Yersinia species labeled wtih horseradish peroxidase. Isolated from a serum pool of rabbits immunized with various strains of heat-killed Yersinia.
Catalog No. 50-669-91