Learn More
Immunology Consultants anti-Goat IgG h+l Antibody (Rabbit) - HRP Conjugated
SDP
Catalog No. 502248875
Encompass_Preferred
RGHL‑50P is an HRP‑conjugated rabbit antibody specific for goat IgG heavy and light chains, designed for chemiluminescent and colorimetric detection. This reagent provides strong, selective binding and robust signal generation in ELISA, Western blotting, and other enzyme‑based assays. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑50P is an excellent choice for high‑sensitivity secondary detection.
This item is not returnable. View return policy
Catalog No. 50-224-8875 Supplier Immunology Consultants Supplier No. RGHL50P
RGHL‑50P is an HRP‑conjugated rabbit antibody specific for goat IgG heavy and light chains, designed for chemiluminescent and colorimetric detection. This reagent provides strong, selective binding and robust signal generation in ELISA, Western blotting, and other enzyme‑based assays. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑50P is an excellent choice for high‑sensitivity secondary detection.