Learn More
Immunology Consultants anti-Guinea Pig IgA Antibody (Sheep) - HRP Conjugated
SDP
Catalog No. 502248855
Encompass_Preferred
This HRP‑conjugated sheep anti‑guinea pig IgA antibody provides sensitive enzymatic detection of IgA in ELISA and Western blotting. The HRP label enables strong chemiluminescent or colorimetric signal generation, supporting high‑sensitivity immunoassays. Supplied at 0.1 mg, it is ideal for small‑scale studies, assay optimization, or diagnostic development. Manufactured by Immunology Consultants, the antibody maintains excellent specificity and low background
This item is not returnable. View return policy
Catalog No. 50-224-8855 Supplier Immunology Consultants Supplier No. SA60PZ
This HRP‑conjugated sheep anti‑guinea pig IgA antibody provides sensitive enzymatic detection of IgA in ELISA and Western blotting. The HRP label enables strong chemiluminescent or colorimetric signal generation, supporting high‑sensitivity immunoassays. Supplied at 0.1 mg, it is ideal for small‑scale studies, assay optimization, or diagnostic development. Manufactured by Immunology Consultants, the antibody maintains excellent specificity and low background