Learn More
Biotium Tyramide Amplification Kit with HRP Streptavidin and CF488A Tyramide
Catalog No. 501964606
Encompass_Preferred
Tyramide amplification, sometimes called Catalyzed Reporter Deposition (CARD), is an enzyme-mediated detection method that uses the catalytic activity of horseradish peroxidase (HRP) to generate high density labeling of a target protein or nucleic acid sequence in situ. The tyramide amplification method has been reported to increase the sensitivity by up to 100-fold compared to the conventional avidin-biotinylated complex procedure. The kit contains HRP Streptavidin CF488A dye tyramide. The dye has an excitation/emission of 490/515 nm. The kit contains sufficient reagent for 50-150 slides.
Catalog No. 50-196-4606 Supplier Biotium Supplier No. 33002
Tyramide amplification, sometimes called Catalyzed Reporter Deposition (CARD), is an enzyme-mediated detection method that uses the catalytic activity of horseradish peroxidase (HRP) to generate high density labeling of a target protein or nucleic acid sequence in situ. The tyramide amplification method has been reported to increase the sensitivity by up to 100-fold compared to the conventional avidin-biotinylated complex procedure. The kit contains HRP Streptavidin CF488A dye tyramide. The dye has an excitation/emission of 490/515 nm. The kit contains sufficient reagent for 50-150 slides.