Learn More
Seracare Life Sciences Inc HRPSTABILIZER
Catalog No. 5067504
Encompass_Preferred
HRP Stabilizer is effective at stabilizing peroxidase conjugates in diluted form for extended periods of time with minimum effect on conjugate activity. Stability studies conducted have indicated the HRPStabilizer retains greater stability of diluted HRP conjugates than 50% glycerol, BSA or other commercially available products when stored at 37 C, room temperature or 4 C. Conjugates may be diluted to a use dilution concentration as low as 1 ug/mL and be stable for at least6 monthswhen stored at 4 C. Product in ready-to-use liquid form.
This item is not returnable. View return policy
Catalog No. 50-675-04 Supplier Seracare Life Sciences Inc Supplier No. 52900005
HRP Stabilizer is effective at stabilizing peroxidase conjugates in diluted form for extended periods of time with minimum effect on conjugate activity. Stability studies conducted have indicated the HRPStabilizer retains greater stability of diluted HRP conjugates than 50% glycerol, BSA or other commercially available products when stored at 37 C, room temperature or 4 C. Conjugates may be diluted to a use dilution concentration as low as 1 ug/mL and be stable for at least6 monthswhen stored at 4 C. Product in ready-to-use liquid form.