Learn More
Aviva Systems Biology Corp RIDA Antibody - N-terminal region (ARP52116_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52116_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HRSP12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HRSP12. Alternate names: P14.5, PSP, UK114ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Rabbit
Catalog No. 50-285-641