Learn More
ACROBiosystems HRSV B Pre-fusion glycoprote
Catalog No. 502534213
Encompass_Preferred
HRSV (B) Pre-fusion glycoprotein F0 His Tag (RSF-V52H8) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Asn 524 (Accession # O36634).
Catalog No. 50-253-4213 Supplier ACROBiosystems Supplier No. RSFV52H850UG
HRSV (B) Pre-fusion glycoprotein F0 His Tag (RSF-V52H8) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Asn 524 (Accession # O36634).